Thermo Scientific Chemicals

Polyvinylpyrrolidone, cross linked, Thermo Scientific Chemicals

Catalog number: 041631.36
500 g, Each
Thermo Scientific Chemicals

Polyvinylpyrrolidone, cross linked, Thermo Scientific Chemicals

Catalog number: 041631.36
500 g, Each
Quantity
Catalog number: 041631.36
also known as 041631-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25249-54-1
Molecular Formula
(C6H9NO)n
SMILES
*-CC(-*)N1CCCC1=O
MDL Number
MFCD00149016
Assay (special)
Nitrogen Content: 11.0-12.8%
Loss on Drying
≤5.0%
Sulfated Ash
≤0.2%
Appearance (Color)
White to off-white
pH
5-8 ( 1% Aqueous solution)

Description

Polyvinylpyrrolidone is used in coatings, printing ink, rubber, glass, leather, cosmetics, soaps, plastics, in paper making and as a stabilizer. It acts as a disintegrant in pharmaceutical tablets and as a fining to extract impurities. It is used to remove polyphenols in beer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text