Thermo Scientific Chemicals
Strontium titanium oxide, S.A. 12.5-25m2/g, Thermo Scientific Chemicals
Catalog number: 041634.22
100 g, Each
Thermo Scientific Chemicals
Strontium titanium oxide, S.A. 12.5-25m2/g, Thermo Scientific Chemicals
Catalog number: 041634.22
100 g, Each
Quantity
Catalog number: 041634.22
also known as 041634-22
Price (USD)
Quantity
-
Have Questions?
Chemical Identifiers
CAS
12060-59-2
Comment
12.5-25m²/g
Comment
0.05-0.15 Micron APS
Description
Strontium titaniumoxide is used as paint . It has been used as a strontium-90-containing material in radioisotope thermoelectric generators. It is also used as a diamond simulant. This compound is been used in television sets, radios and computers, capacitors, because of the dielectric constant of the material. Used in electronics and in electrical insulation.