Thermo Scientific Chemicals

Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 041665.18
50 g, Each
Thermo Scientific Chemicals

Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 041665.18
50 g, Each
Quantity
Catalog number: 041665.18
also known as 041665-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12070-13-2
IUPAC Name
methyliumylidyneditungstenene
Molecular Formula
CW2
InChI Key
FHLFYDOXSZQEPO-UHFFFAOYSA-N
SMILES
[W-]=[W]#[C+]
Total Metal Impurities
0.5% max.

Description

Tungsten carbide is used in industrial machinery, cutting tools, abrasives, armor-piercing rounds and jewelry. It is preferred even over stainless steel because of their incredible hardness and resistance to wear and tear. It is utilized for the manufacture of mill products including grinding and milling. It also includes tipping trekking, ski poles as well as cleats. It is mainly used in the form of cemented tungsten carbide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text