Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals
Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 041665.30
also known as 041665-30
Price (USD)/ Each
640.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
640.00
Add to cart
Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number041665.30
Price (USD)/ Each
640.00
-
Add to cart
Chemical Identifiers
CAS12070-13-2
IUPAC Namemethyliumylidyneditungstenene
Molecular FormulaCW2
InChI KeyFHLFYDOXSZQEPO-UHFFFAOYSA-N
SMILES[W-]=[W]#[C+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Tungsten carbide is used in industrial machinery, cutting tools, abrasives, armor-piercing rounds and jewelry. It is preferred even over stainless steel because of their incredible hardness and resistance to wear and tear. It is utilized for the manufacture of mill products including grinding and milling. It also includes tipping trekking, ski poles as well as cleats. It is mainly used in the form of cemented tungsten carbide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only