Bismuth titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Bismuth titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 041669.30
also known as 041669-30
Price (USD)/ Each
360.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
360.00
Add to cart
Bismuth titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number041669.30
Price (USD)/ Each
360.00
-
Add to cart
Chemical Identifiers
CAS12233-34-0
IUPAC Nametetraoxo-2,4,6,8,10-pentaoxa-1,11-dibisma-3,5,7,9-tetratitanaundecane-1,11-dione
Molecular FormulaBi2O11Ti4
InChI KeyTZQKEPWGUIJJIY-UHFFFAOYSA-N
SMILESO=[Bi]O[Ti](=O)O[Ti](=O)O[Ti](=O)O[Ti](=O)O[Bi]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only