Thermo Scientific™

Triisopropylphosphine, 98%, Thermo Scientific Chemicals

Catalog number: 041684.09
10 g, Each
Thermo Scientific™

Triisopropylphosphine, 98%, Thermo Scientific Chemicals

Catalog number: 041684.09
10 g, Each
Quantity
Catalog number: 041684.09
also known as 041684-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6476-36-4
IUPAC Name
tris(propan-2-yl)phosphane
Molecular Formula
C9H21P
InChI Key
IGNTWNVBGLNYDV-UHFFFAOYSA-N
SMILES
CC(C)P(C(C)C)C(C)C
Appearance (Color)
Colorless
Form
Liquid
Phosphorus-31 NMR
> 97.0%

Description

Triisopropylphosphine is used as a ligand in organometallic chemistry. It is also used to produce chlor(triisopropyl)phosphonium- dimesylamid at temperature of 40°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text