Thermo Scientific Chemicals

Poly(styrene sulfonic acid) sodium salt, M.W. 70,000, Thermo Scientific Chemicals

Catalog number: 041688.18
50 g, Each
Thermo Scientific Chemicals

Poly(styrene sulfonic acid) sodium salt, M.W. 70,000, Thermo Scientific Chemicals

Catalog number: 041688.18
50 g, Each
Quantity
Catalog number: 041688.18
also known as 041688-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25704-18-1
Molecular Formula
(C8H7NaO3S)n
SMILES
[Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*
MDL Number
MFCD00084449
Appearance (Color)
White to cream to pale brown
Form
Powder
Comment
Specification differs for U.S. and non-U.S. material where indicated
Infrared spectrum
Conforms
Volatiles
≤12.0% (U.S. specification)

Description

Poly(styrene sulfonic acid) sodium salt is used as Polyelectrolyte. It is also used as electroconductive and antistatic resin for electrographic and electrophotographic substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text