Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals
Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 041690.30
also known as 041690-30
Price (USD)/ Each
143.65
Online exclusive
159.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
143.65
Online exclusive
159.00 
Save 15.35 (10%)
Add to cart
Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals
Catalog number041690.30
Price (USD)/ Each
143.65
Online exclusive
159.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2695-37-6
IUPAC Namesodium 4-ethenylbenzene-1-sulfonate
Molecular FormulaC8H7NaO3S
InChI KeyXFTALRAZSCGSKN-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale brown
Assay from Supplier's CofA≥79%
Styrene-4-sulfonic acid sodium salt is grafted on poly(ethersulfone) membrane to determine the fractionation of proteins with similar molecular weights. It is used as sodium based ion exchange membranes for membrane capacitive deionization (MCDI) for synthesis. It is used in the study of copolymerization characteristics of sodium styrenesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only