Multi Ion anion IC standard solution in H2O, Specpure™, Thermo Scientific Chemicals
Multi Ion anion IC standard solution in H<sub>2</sub>O, Specpure&trade;, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Multi Ion anion IC standard solution in H2O, Specpure™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Catalog number 041693.AE
also known as 041693-AE
Price (USD)/ Each
237.65
Online exclusive
264.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
237.65
Online exclusive
264.00 
Save 26.35 (10%)
Add to cart
Multi Ion anion IC standard solution in H2O, Specpure™, Thermo Scientific Chemicals
Catalog number041693.AE
Price (USD)/ Each
237.65
Online exclusive
264.00 
Save 26.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialMulti Ion anion IC standard solution in H2O
Name NoteBr-, Cl-, F-, NO3-, PO4-3 and SO4-2at 100μg/mL. Keep refrigerated
Quantity100 mL
Recommended StorageAmbient temperatures
EINECS Number231-791-2
View more
Multi Ion anion IC standard solution in H2O is utilized as a standard solution in analytical chemistry and ion chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only