Multi Ion cation IC standard solution, Specpure™ (Dionex) in 5% HNO{3}, Thermo Scientific Chemicals
Multi Ion cation IC standard solution, Specpure™ (Dionex) in 5% HNO{3}, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Multi Ion cation IC standard solution, Specpure™ (Dionex) in 5% HNO{3}, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Catalog number 041695.AE
also known as 041695-AE
Price (USD)/ Each
234.65
Online exclusive
261.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
234.65
Online exclusive
261.00 
Save 26.35 (10%)
Add to cart
Multi Ion cation IC standard solution, Specpure™ (Dionex) in 5% HNO{3}, Thermo Scientific Chemicals
Catalog number041695.AE
Price (USD)/ Each
234.65
Online exclusive
261.00 
Save 26.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialMulti Ion cation IC standard solution
Concentration(Dionex) in 5% HNO3
Health Hazard 1H290-H314-H335
Quantity100 mL
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
View more
Multi Ion cation IC standard solution is a high purity starting material, which is a certified primary reference material for analytical and Ion Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only