Thermo Scientific Chemicals

Arsenic(III) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 041709.18
50 g, Each
Thermo Scientific Chemicals

Arsenic(III) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 041709.18
50 g, Each
Quantity
Catalog number: 041709.18
also known as 041709-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12044-54-1
IUPAC Name
diarsenic tritellurium
Molecular Formula
As2Te3
InChI Key
KOOQXSDVIQPGQD-UHFFFAOYSA-N
SMILES
[As].[As].[Te].[Te].[Te]
Total Metal Impurities
0.001% max.

Description

Vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text