Ethylaluminum dichloride, 1M in hexanes, Thermo Scientific Chemicals
Ethylaluminum dichloride, 1M in hexanes, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylaluminum dichloride, 1M in hexanes, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
250 mL
Catalog number 041712.AB
also known as 041712-AB
Price (USD)/ Each
17.40
-
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (USD)/ Each
17.40
Add to cart
Ethylaluminum dichloride, 1M in hexanes, Thermo Scientific Chemicals
Catalog number041712.AB
Price (USD)/ Each
17.40
-
Add to cart
Chemical Identifiers
CAS563-43-9
IUPAC Namedichloro(ethyl)alumane
Molecular FormulaC2H5AlCl2
InChI KeyUAIZDWNSWGTKFZ-UHFFFAOYSA-L
SMILESCC[Al](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Concentration0.95-1.10M
Ethylaluminum dichloride is used as a catalyst component in the polyolefin industry, for the synthesis of synthetic rubbers and for the dimerisation of olefins. It is also employed as a catalyst for Friedel-Crafts acylations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only