Thermo Scientific Chemicals

Sodium diphenylamine-4-sulfonate, ACS, Thermo Scientific Chemicals

Catalog number: 041714.18
50 g, Each
Thermo Scientific Chemicals

Sodium diphenylamine-4-sulfonate, ACS, Thermo Scientific Chemicals

Catalog number: 041714.18
50 g, Each
Quantity
Catalog number: 041714.18
also known as 041714-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6152-67-6
IUPAC Name
4-(phenylamino)benzene-1-sulfonic acid sodium
Molecular Formula
C12H11NNaO3S
InChI Key
VMVSDVIYIMNZBS-UHFFFAOYSA-N
SMILES
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Form
powder
Appearance (Color)
Off-white to grey
Comment
Sensitivity as indicator P.T.

Description

Redox indicatorSodium diphenylamine-4-sulfonate is used as a pH independent redox indicator used in the determination of redox potential. It is used in the preparation of poly-(aninile-co-N-(4-sulfophenyl)aniline) by polymerization with aniline. It acts as a ligand and form complexes with metal halides like lanthanum(III) chloride. It is also involved in the synthesis of gold nano crystals by reaction with chloroauric acid in the presence of polyethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text