Thermo Scientific Chemicals

Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate, Thermo Scientific Chemicals

Catalog number: 041715.22
100 g, Each
Thermo Scientific Chemicals

Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate, Thermo Scientific Chemicals

Catalog number: 041715.22
100 g, Each
Quantity
Catalog number: 041715.22
also known as 041715-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123409-01-8
IUPAC Name
naphthalene-1,3,6-trisulfonic acid hydrate trisodium
Molecular Formula
C10H10Na3O10S3
InChI Key
BMCNZRJPDRPEDI-UHFFFAOYSA-N
SMILES
O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
Assay from Suppliers CofA
≥95.0% (ion exchange titration, anhydrous basis)
Assay from Suppliers CofA
≥65.0% (HPLC)
Water Content (Karl Fischer Titration)
≤12.0%
Appearance (Color)
White to yellow to pale brown
Form
Crystals or powder or crystalline powder

Description

Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate is used as an anionic chromophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text