Thermo Scientific Chemicals

Calcium boride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 041722.22
100 g, Each
Thermo Scientific Chemicals

Calcium boride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 041722.22
100 g, Each
Quantity
Catalog number: 041722.22
also known as 041722-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12007-99-7
IUPAC Name
calcium pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide
Molecular Formula
B6Ca
InChI Key
AWVIDOFQNLOVRC-UHFFFAOYSA-N
SMILES
[Ca++].B12B3[B-]11B4B1[B-]234
Form
Powder, -325 mesh
Total Metal Impurities
≤0.5%

Description

Calcium boride is used as an electrode (cathode) material and n-type thermoelectric materials. It is used to make boron- alloyed steel, as a deoxidation agent in the production of oxygen-free copper. It is further used as an antioxidant in carbon bonded refractories. It acts as a new material which is used in the nuclear-industry for neutron- preventing, high-purity metal borid (TiB2,ZrB2,HfB2) and high-purity boron alloy(Ni-B, Co-B, Cu-b).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text