Thermo Scientific Chemicals

n-Hexane, Spectrophotometric Grade, 95+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041727.AH
200 mL, Each
Thermo Scientific Chemicals

n-Hexane, Spectrophotometric Grade, 95+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041727.AH
200 mL, Each
Quantity
Catalog number: 041727.AH
also known as 041727-AH
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-54-3
IUPAC Name
hexane
Molecular Formula
C6H14
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
Water Content (Karl Fischer Titration)
0.02%, max
Form
clear liquid
Optical Absorbance at
250 nm: 0.01 max, 220 nm 0.10 max, 210 nm 0.30 max, 195 nm 1.00 max
Appearance (Color)
Colorless
Assay (GC)
> 95.0%

Description

n-Hexane is used in the formulation of glues for shoes, leather products, and roofing, as a cleaning agent, de-greasing agent. It is used to extract edible oils from seeds and vegetables, a non-polar solvent in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text