Thermo Scientific Chemicals

Lithium iodide, anhydrous, 98+%, Thermo Scientific Chemicals

Catalog number: 041733.18
50 g, Each
Thermo Scientific Chemicals

Lithium iodide, anhydrous, 98+%, Thermo Scientific Chemicals

Catalog number: 041733.18
50 g, Each
Quantity
Catalog number: 041733.18
also known as 041733-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10377-51-2
IUPAC Name
lithium(1+) iodide
Molecular Formula
ILi
InChI Key
HSZCZNFXUDYRKD-UHFFFAOYSA-M
SMILES
[Li+].[I-]
Total Metal Impurities
≤ 2.0%
Form
Lumps/powder

Description

Lithium iodide is used as reagent in the cleavage of esters, ethers and epoxides, and carboxylation. It is used in scintillation counters. Lithium iodide is used as an electrolyte in high temperature batteries and artificial pacemakers. Phosphor, which is a complex of iodine, is used in neuron detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text