Thermo Scientific Chemicals

Di-n-octyldichlorotin, 95%, Thermo Scientific Chemicals

Catalog number: 041738.06
5 g, Each
Thermo Scientific Chemicals

Di-n-octyldichlorotin, 95%, Thermo Scientific Chemicals

Catalog number: 041738.06
5 g, Each
Quantity
Catalog number: 041738.06
also known as 041738-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3542-36-7
Molecular Formula
C16H36Cl2Sn
InChI Key
DXPJPPSKHFECLR-UHFFFAOYSA-N
SMILES
Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC
Molecular Weight (g/mol)
418.07
Appearance (Color)
Pale yellow to orange
Assay from Supplier's CofA
≥94%
Form
Crystalline solid
Melting Point
44 -48?C

Description

Di-n-octyldichlorotin compounds are used as intermediates in the production of other organotin chemicals. Metatin and Acima. These products are also used as catalysts or industrial intermediates in the production of polyurethanes, silicone resins, synthetic lubricants, and MS polymer sealants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text