Thermo Scientific Chemicals

Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals

Catalog number: 041748.22
100 g, Each
Thermo Scientific Chemicals

Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals

Catalog number: 041748.22
100 g, Each
Quantity
Catalog number: 041748.22
also known as 041748-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68131-04-4
IUPAC Name
disodium bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
Molecular Formula
C9H8Na2O4
InChI Key
HMGUIQPKFUZDPV-UHFFFAOYNA-L
SMILES
[Na+].[Na+].[O-]C(=O)C1C2CC(C=C2)C1C([O-])=O
Assay (unspecified)
45 to 70% ( as Humic acid)
pH
9-10
Loss on Drying
15% max

Description

Humic acid sodium salt is a principal component of humic substances. It boosts plant growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text