Phthalic acid, ACS, 99.5+%, Thermo Scientific Chemicals
Phthalic acid, ACS, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phthalic acid, ACS, 99.5+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 041770.36
also known as 041770-36
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
Add to cart
Phthalic acid, ACS, 99.5+%, Thermo Scientific Chemicals
Catalog number041770.36
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS88-99-3
IUPAC Namebenzene-1,2-dicarboxylic acid
Molecular FormulaC8H6O4
InChI KeyXNGIFLGASWRNHJ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCl 0.001%
Impurity contentNO₃ 0.005%
Insoluble material0.05%
Residual water0.5%
Impurity contentFe 0.001%
View more
Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only