Thermo Scientific Chemicals

Phthalic anhydride, ACS, 99.0-100.2%, Thermo Scientific Chemicals

Catalog number: 041771.36
500 g, Each
Thermo Scientific Chemicals

Phthalic anhydride, ACS, 99.0-100.2%, Thermo Scientific Chemicals

Catalog number: 041771.36
500 g, Each
Quantity
Catalog number: 041771.36
also known as 041771-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85-44-9
IUPAC Name
1,3-dihydro-2-benzofuran-1,3-dione
Molecular Formula
C8H4O3
InChI Key
LGRFSURHDFAFJT-UHFFFAOYSA-N
SMILES
O=C1OC(=O)C2=CC=CC=C12
Residue on ignition
0.01%
Impurity content
Cl 0.002%
Impurity content
Fe 5ppm
Impurity content
SO₄ 0.003%
Melting Point
not more than 3? range including 131?

Description

Phthalic anhydride is used as an intermediate in chemical synthesis and plastics manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text