Thermo Scientific™

Sodium telluride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 041777.09
10 g, Each
Thermo Scientific™

Sodium telluride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 041777.09
10 g, Each
Quantity
Catalog number: 041777.09
also known as 041777-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12034-41-2
IUPAC Name
disodium tellanediide
Molecular Formula
Na2Te
InChI Key
QZBKIBAHGRAIOX-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[Te--]
Total Metal Impurities
0.1% max.

Description

Sodium telluride is used as a semiconductor, as a reducing agent and to prepare organic tellurium compounds. It is used in the reduction of aromatic carbonyl compounds to alcohols. It is also used in the preparation of pyrrolo[2,3-d]pyrimidines (7-deaza-9H-purines) from aromatic nitriles. It is used to prepare nanocrystalline zinc telluride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text