Thermo Scientific Chemicals

Zinc acrylate, Thermo Scientific Chemicals

Catalog number: 041780.A1
1 kg, Each
Thermo Scientific Chemicals

Zinc acrylate, Thermo Scientific Chemicals

Catalog number: 041780.A1
1 kg, Each
Quantity
Catalog number: 041780.A1
also known as 041780-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14643-87-9
IUPAC Name
zinc(2+) bis(prop-2-enoate)
Molecular Formula
C6H6O4Zn
InChI Key
XKMZOFXGLBYJLS-UHFFFAOYSA-L
SMILES
[Zn++].[O-]C(=O)C=C.[O-]C(=O)C=C
Appearance (Color)
White
Form
Crystalline powder
Assay from Supplier's CofA
≥94%
Melting Point
> 250?C

Description

The primary use of Zinc Acrylate is in the production of golf balls. Other uses are primarily in the production of specialty rubber products such as belts and hoses for automobiles. Also used as a tissue adhesive used for adherence of tissue to tissue or tissue to non-tissue for prostheses in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text