Thermo Scientific Chemicals

Boron oxide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 041786.09
10 g, Each
Thermo Scientific Chemicals

Boron oxide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 041786.09
10 g, Each
Quantity
Catalog number: 041786.09
also known as 041786-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1303-86-2
IUPAC Name
oxo[(oxoboranyl)oxy]borane
Molecular Formula
B2O3
InChI Key
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
SMILES
O=BOB=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay from Supplier's CofA
≤0.001% (≤10ppm) Total metal Impurities (Emission Spectroscopy)

Description

Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants and dopants in semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text