Thermo Scientific Chemicals

Tungsten(V) ethoxide, Thermo Scientific Chemicals

Catalog number: 041794.09
10 g, Each
Thermo Scientific Chemicals

Tungsten(V) ethoxide, Thermo Scientific Chemicals

Catalog number: 041794.09
10 g, Each
Quantity
Catalog number: 041794.09
also known as 041794-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26143-11-3
IUPAC Name
pentaethoxytungsten
Molecular Formula
C10H25O5W
InChI Key
SRGJJDPFERQMIL-UHFFFAOYSA-N
SMILES
CCO[W](OCC)(OCC)(OCC)OCC
Specific Gravity
1.44 to 1.47
Appearance (Color)
Clear to hazy, colorless to yellow to purple
Form
Liquid

Description

Tungsten(V) ethoxide is used as an intermediate for sol-gel coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text