Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh, Thermo Scientific Chemicals
Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh, Thermo Scientific Chemicals

For the preparation of functionalized beads used in solid-phase synthesis
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 041800.22
also known as 041800-22
Price (USD)/ Each
278.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
278.00
Add to cart
Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh, Thermo Scientific Chemicals
Catalog number041800.22
Price (USD)/ Each
278.00
-
Add to cart
Specifications
Chemical Name or MaterialPoly(styrene-divinylbenzene)
Name Note1% cross-linked
CAS9003-70-7
Physical FormBeads
Recommended StorageAmbient temperatures
View more
Poly(styrene-divinylbenzene) is widely used as column packing material in liquid chromatography for separation of both organic and inorganic ions. is used in inorganic trace analysis for solid phase extraction preconcentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only