Platinum Iridium foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals
Platinum Iridium foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum Iridium foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 041803.FF
Price (USD)/ Each
280.00
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
280.00
Add to cart
Platinum Iridium foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals
Catalog number041803.FF
Price (USD)/ Each
280.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPlatinum Iridium foil
Name Note0.05mm (0.002 in.) thick
Recommended StorageAmbient temperatures
Avg. Mol. Wt. or Mol. Wt. RangePt:Ir; 90:10 wt%
TSCAYes
View more
Platinum Iridium foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding precious metals) is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.