Thermo Scientific Chemicals
Platinum Rhodium foil, 0.05mm (0.002 in.) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 041809.FI
50 x 50 mm, Each
Thermo Scientific Chemicals
Platinum Rhodium foil, 0.05mm (0.002 in.) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 041809.FI
50 x 50 mm, Each
Quantity
Catalog number: 041809.FI
also known as 041809-FI
Price (USD)
Have Questions?
Description
Platinum Rhodium foil, 0.05mm (0.002in) thick, 99.9% (metals basis) is used in the glass industry for furnace linings and complex fabrications. It is also used as a plating material. Further, it acts as a catalyst and as a standard for calibration of base metal thermocouples.