Platinum Rhodium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum Rhodium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum Rhodium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Used as a standard for calibration of base metal thermocouples. It is also used in the glass industry for furnace linings and complex fabrications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
Catalog number 041811.GH
also known as 041811-GH
Price (USD)/ Each
-
Quantity:
100 x 100 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum Rhodium foil
Recommended StorageAmbient temperatures
Quantity100 x 100 mm
TSCAYes
Molecular FormulaPt:Rh; 80:20 wt%
View more
Platinum Rhodium foil is used as a standard for calibration of base metal thermocouples. It is also used in the glass industry for furnace linings and complex fabrications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only