Thermo Scientific Chemicals

Platinum gauze, 45 mesh woven from 0.198mm (0.0078in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 041814.FI
50 x 50 mm, Each
Thermo Scientific Chemicals

Platinum gauze, 45 mesh woven from 0.198mm (0.0078in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 041814.FI
50 x 50 mm, Each
Quantity
Catalog number: 041814.FI
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-06-4
IUPAC Name
platinum
Molecular Formula
Pt
InChI Key
BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES
[Pt]
Appearance (Color)
Metallic grey
Form
Gauze, 45 mesh, 0.0078 inch wire dia.

Description

Platinum gauze is used in electronics industry for electrical contacts which might be subjected to high temperatures. It also acts as a catalyst in the production of nitric acid. It plays an essential role in the cracking processes in oil refineries. It can be coated on electrode surface by electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text