Tetrahydrofuran, Spectrophotometric grade, 99.7+%, unstab., packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric grade, 99.7+%, unstab., packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific™

Tetrahydrofuran, Spectrophotometric grade, 99.7+%, unstab., packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 x 1 L
Catalog number 041819.AK
also known as 041819-AK
Price (USD)/ Each
40.65
Online exclusive
44.60 
Save 3.95 (9%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
40.65
Online exclusive
44.60 
Save 3.95 (9%)
Add to cart
Tetrahydrofuran, Spectrophotometric grade, 99.7+%, unstab., packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number041819.AK
Price (USD)/ Each
40.65
Online exclusive
44.60 
Save 3.95 (9%)
-
Add to cart
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation≤5.0 ppm
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay from Supplier's CofA>99.7%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only