Thermo Scientific Chemicals

Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm, Thermo Scientific Chemicals

Catalog number: 041826.22
100 g, Each
Thermo Scientific Chemicals

Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm, Thermo Scientific Chemicals

Catalog number: 041826.22
100 g, Each
Quantity
Catalog number: 041826.22
also known as 041826-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14177-51-6
IUPAC Name
nickel(2+) dioxotungstenbis(olate)
Molecular Formula
NiO4W
InChI Key
QLTKZXWDJGMCAR-UHFFFAOYSA-N
SMILES
[Ni++].[O-][W]([O-])(=O)=O
Impurity content
Co <100ppm
Total Metal Impurities
0.1% max.

Description

Nickel tungsten oxide is used as photo catalysts, sensors and humidity dielectric resonators. It is considered as a new cathode material for asymmetric super capacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text