Copper(I) chloride, 99% (metals basis), Thermo Scientific Chemicals
Copper(I) chloride, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(I) chloride, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 041837.36
also known as 041837-36
Price (USD)/ Each
413.65
Online exclusive
459.00 
Save 45.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
413.65
Online exclusive
459.00 
Save 45.35 (10%)
Add to cart
Copper(I) chloride, 99% (metals basis), Thermo Scientific Chemicals
Catalog number041837.36
Price (USD)/ Each
413.65
Online exclusive
459.00 
Save 45.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7758-89-6
IUPAC Nameλ¹-copper(1+) chloride
Molecular FormulaClCu
InChI KeyOXBLHERUFWYNTN-UHFFFAOYSA-M
SMILES[Cl-].[Cu+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Copper(I) chloride is precursor to many copper compounds including copper oxychloride and many organocuprate compounds of synthetic interest. It is used as a catalyst as well as a reagent in many organic reactions including Gatterman-Koch, Sandmeyer, Grignard and Gilman reactions. It catalyzes 1,4-addition of Grignard reagents to alpha,beta-unsaturated ketones. It is also used as a doping salt to enhance the cationization of polystyrene in matrix-assisted laser desorption/ionization mass spectrometry. It is also an intermediate formed in the Wacker process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only