tert-Butyl methyl ether, HPLC grade, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
tert-Butyl methyl ether, HPLC grade, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl methyl ether, HPLC grade, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Chromatography solvent
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 041839.AK
also known as 041839-AK
Price (USD)/ Each
35.65
Online exclusive
39.00 
Save 3.35 (9%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
35.65
Online exclusive
39.00 
Save 3.35 (9%)
Add to cart
tert-Butyl methyl ether, HPLC grade, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number041839.AK
Price (USD)/ Each
35.65
Online exclusive
39.00 
Save 3.35 (9%)
-
Add to cart
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at225 nm: ≤0.50 AU
Peroxide content≤0.0001% (as H₂O₂ at time of packaging)
Color (APHA)≤10
Optical Absorbance at210 nm: ≤1.0 AU
Optical Absorbance at250 nm: ≤0.10 AU
View more
tert-Butyl methyl ether is used as a solvent for chromatography and in the manufacturing of chemicals, drugs, and drug intermediates. It is used as a mobile phase in High Performance Liquid Chromatography. It acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline (oxygenates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only