Thermo Scientific Chemicals

1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041842.K2
1 L, Each
Thermo Scientific Chemicals

1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041842.K2
1 L, Each
Quantity
Catalog number: 041842.K2
also known as 041842-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-23-8
IUPAC Name
propan-1-ol
Molecular Formula
C3H8O
InChI Key
BDERNNFJNOPAEC-UHFFFAOYSA-N
SMILES
CCCO
Water Content (Karl Fischer Titration)
≤50.0 ppm
Appearance (Color)
Clear colorless
Assay from Supplier's CofA
≥99.9%
Form
Liquid
Residue on Evaporation
≤3.0 ppm

Description

Used in making cosmetics, skin and hair preparations, pharmaceuticals, perfumes, lacquer formulations, dye solutions, antifreezes, rubbing alcohols, soaps, window cleaners, acetone and other chemicals and products. Also referred as ropyl alcohol;n-Propan-1-ol;n-Propanol;n-Propyl alcohol, Ethylcarbinol, Optal, Osmosol extra.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text