1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
250 mL
1 L
Catalog number 041842.M1
also known as 041842-M1
Price (USD)/ Each
262.00
-
Add to cart
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)/ Each
262.00
Add to cart
1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number041842.M1
Price (USD)/ Each
262.00
-
Add to cart
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤50.0 ppm
Appearance (Color)Clear colorless
Assay from Supplier's CofA≥99.9%
FormLiquid
Residue on Evaporation≤3.0 ppm
Used in making cosmetics, skin and hair preparations, pharmaceuticals, perfumes, lacquer formulations, dye solutions, antifreezes, rubbing alcohols, soaps, window cleaners, acetone and other chemicals and products. Also referred as ropyl alcohol;n-Propan-1-ol;n-Propanol;n-Propyl alcohol, Ethylcarbinol, Optal, Osmosol extra.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only