N,N-Dimethylformamide, HPLC grade, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
N,N-Dimethylformamide, HPLC grade, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylformamide, HPLC grade, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Same HPLC specifications as 22915
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number 041859.K2
also known as 041859-K2
Price (USD)/ Each
76.65
Online exclusive
84.90 
Save 8.25 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
76.65
Online exclusive
84.90 
Save 8.25 (10%)
Add to cart
N,N-Dimethylformamide, HPLC grade, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number041859.K2
Price (USD)/ Each
76.65
Online exclusive
84.90 
Save 8.25 (10%)
-
Add to cart
Chemical Identifiers
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
UV/VisAt 270 nm 1.00 AU, max
UV/VisAt 295 nm 0.10 AU, max
UV/VisAt 340 nm 0.01 AU, max
Residue on Evaporation5 ppm, max
View more
N,N-Dimethylformamide is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only