Thermo Scientific Chemicals

(1-Hexadecyl)trimethylammonium chloride, 96%, Thermo Scientific Chemicals

Catalog number: 041861.36
500 g, Each
Thermo Scientific Chemicals

(1-Hexadecyl)trimethylammonium chloride, 96%, Thermo Scientific Chemicals

Catalog number: 041861.36
500 g, Each
Quantity
Catalog number: 041861.36
also known as 041861-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-02-7
IUPAC Name
hexadecyltrimethylazanium chloride
Molecular Formula
C19H42ClN
InChI Key
WOWHHFRSBJGXCM-UHFFFAOYSA-M
SMILES
[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C
Assay (Titration)
> 95.0%
Appearance (Color)
White to off-white
Form
powder

Description

(1-Hexadecyl)trimethylammonium chloride is used as a topical antiseptic and surfactant. It is used in hair care formulations, cream rinse ingredient, hair conditioners and shampoos as a conditioning agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text