Thermo Scientific Chemicals

Iron(III) ethoxide, 99.6% (metals basis), Thermo Scientific Chemicals

Catalog number: 041863.18
50 g, Each
Thermo Scientific Chemicals

Iron(III) ethoxide, 99.6% (metals basis), Thermo Scientific Chemicals

Catalog number: 041863.18
50 g, Each
Quantity
Catalog number: 041863.18
also known as 041863-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5058-42-4
IUPAC Name
iron(3+) triethanolate
Molecular Formula
C6H15FeO3
InChI Key
JTPUGUWXHGEEHW-UHFFFAOYSA-N
SMILES
[Fe+3].CC[O-].CC[O-].CC[O-]
Total Metal Impurities
0.4% max.

Description

Precursor for Sol-Gel processes.Iron(III) ethoxide is used as a precursor for inorganic compound iron oxide. It is used in the preparation of iron-platinum nanoparticles by pyrolysis with platinum(II) acetylacetonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text