Thermo Scientific Chemicals

1-Butanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041867.M1
4 x 1 L, Each
Thermo Scientific Chemicals

1-Butanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041867.M1
4 x 1 L, Each
Quantity
Catalog number: 041867.M1
also known as 041867-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-36-3
IUPAC Name
butan-1-ol
Molecular Formula
C4H10O
InChI Key
LRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILES
CCCCO
Residue on Evaporation
≤ 10ppm
Form
Liquid, packaged under Argon in resealable ChemSeal? bottles.
Appearance (Color)
Clear colorless, ≤10 APHA.
Assay (GC)
GC-FID ≥99.9 % (corrected for water)
Water Content (Karl Fischer Titration)
≤ 50ppm

Description

1-Butanol is used as solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text