Thermo Scientific Chemicals

2-Butanol, anhydrous, 99%, packaged under Argon in resealable ChemSeal bottles, Thermo Scientific Chemicals

Catalog number: 041871.K2
1 L, Each
Thermo Scientific Chemicals

2-Butanol, anhydrous, 99%, packaged under Argon in resealable ChemSeal bottles, Thermo Scientific Chemicals

Catalog number: 041871.K2
1 L, Each
Quantity
Catalog number: 041871.K2
also known as 041871-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-92-2
IUPAC Name
butan-2-ol
Molecular Formula
C4H10O
InChI Key
BTANRVKWQNVYAZ-UHFFFAOYNA-N
SMILES
CCC(C)O
Appearance (Color)
Clear, colorless
Form
liquid
Assay (GC)
> 98.5%
Water Content (Karl Fischer Titration)
< 100 ppm

Description

2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text