Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 0.9-1.1M in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041872.36
500 g, Each
Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 0.9-1.1M in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041872.36
500 g, Each
Quantity
Catalog number: 041872.36
also known as 041872-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4039-32-1
IUPAC Name
lithium(1+) bis(trimethylsilyl)azanide
Molecular Formula
C6H18LiNSi2
InChI Key
YNESATAKKCNGOF-UHFFFAOYSA-N
SMILES
[Li+].C[Si](C)(C)[N-][Si](C)(C)C
Appearance (Color)
Colorless to pale yellow
Form
Liquid
Concentration
0.90-1.10M

Description

Lithium bis(trimetylsilyl)amide is used as a strong base in organic synthesis and as catalysts in polymerization reactions. It is also used to form various organolithium compounds such as acetylides and lithium enolates. It is a base employed in generating enolates for preparation of lactone precursors, pyranones and cyclohexanes. Further, it is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines. In addition, it is used in a three-step synthesis of disubstituted 1,2,5-thiadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text