Chemical Name or MaterialSulfur in Crude Oil standard solution
Name Note40,000μg/g (4.00%)
Health Hazard 1H226-H335-H350
Quantity100 mL
Health Hazard 2GHS H Statement H350-H226 May cause cancer. Flammable liquid and vapor.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.