Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyimide film, 0.008mm thick, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
300 x 300 mm
100 x 100 mm
150 x 150 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041896.HB
Price (USD)/ Each
-
Quantity:
300 x 300 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyimide film
Name Note0.008mm thick
CAS62929-02-6
Physical Form≈0.1136g/100x100mm
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only