2,2,4-Trimethylpentane, Environmental Grade, 99.5+%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Environmental Grade, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, Environmental Grade, 99.5+%, Thermo Scientific Chemicals

Suitable for pesticide residue analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 4 L
Catalog number 041904.K2
also known as 041904-K2
Price (USD)/ Each
57.20
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
57.20
Add to cart
2,2,4-Trimethylpentane, Environmental Grade, 99.5+%, Thermo Scientific Chemicals
Catalog number041904.K2
Price (USD)/ Each
57.20
-
Add to cart
Chemical Identifiers
CAS540-84-1
IUPAC Name2,2,4-trimethylpentane
Molecular FormulaC8H18
InChI KeyNHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILESCC(C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)10
Residual water0.02%Note: Filtered through 0.2? filters
Residual halide5ppt (GC-ECD)
Residue on Evaporation3ppm
Suitable for pesticide residue analysis2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only