Ethylene glycol diacetate, 97%, Thermo Scientific Chemicals
Ethylene glycol diacetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylene glycol diacetate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number 041910.0E
also known as 041910-0E
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
Add to cart
Ethylene glycol diacetate, 97%, Thermo Scientific Chemicals
Catalog number041910.0E
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS111-55-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (GC)≥96.0%
Water Content (Karl Fischer Titration)≤0.2%
Refractive Index1.412 to 1.416
FormLiquid
EGDA imparts excellent flow properties in baking lacquers and enamels and where thermoplastic acrylic resins are used. It is also a good solvent for cellulosic coatings and can be used in some ink systems such as screen inks. It has found use as a perfume fixative, and has reported applications in waterborne adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only