Copper Beryllium foil, alloy C17200, 0.25mm (0.01in) thick, Thermo Scientific Chemicals
Copper Beryllium foil, alloy C17200, 0.25mm (0.01in) thick, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper Beryllium foil, alloy C17200, 0.25mm (0.01in) thick, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 x 200 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041913.GU
Price (USD)/ Each
-
Quantity:
50 x 200 mm
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialCopper Beryllium foil, Alloy C17200
Name Note0.25mm (0.01 in.) thick
Health Hazard 1H317-H332-H335-H350i-H373
Health Hazard 2GHS H Statement
H350-H373-H332-H317
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c
View more
Uses include springs of all kinds where high strength and corrosion resistance are important, resistance welding electrodes, dies and high strength, corrosion resistant tubing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only