Molybdenum powder, spherical, -170 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Molybdenum powder, spherical, -170 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum powder, spherical, -170 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041919.30
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSpherical powder
Elemental AnalysisMo > 99.5%
Particle Size+170 mesh 1.0% max
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only