Magnesium citrate hydrate, Mg ca 16% (dry wt.), Thermo Scientific Chemicals
Magnesium citrate hydrate, Mg ca 16% (dry wt.), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium citrate hydrate, Mg ca 16% (dry wt.), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 041921.A1
also known as 041921-A1
Price (USD)/ Each
52.65
Online exclusive
57.90 
Save 5.25 (9%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
52.65
Online exclusive
57.90 
Save 5.25 (9%)
Add to cart
Magnesium citrate hydrate, Mg ca 16% (dry wt.), Thermo Scientific Chemicals
Catalog number041921.A1
Price (USD)/ Each
52.65
Online exclusive
57.90 
Save 5.25 (9%)
-
Add to cart
Chemical Identifiers
CAS960307-84-0
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals≤10ppm
Impurity contentCalcium, Ca: ≤0.15%
Impurity contentLead, Pb: ≤0.5ppm
Appearance (Color)White to pale cream
Assay from Suppliers CofAMagnesium, Mg: ≥14.5 to ≤16.4%
View more
Used as a laxative or dietary supplement

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only