Thermo Scientific Chemicals

Magnesium nitride, 99.6% (metals basis), Thermo Scientific Chemicals

Catalog number: 041946.18
50 g, Each
Thermo Scientific Chemicals

Magnesium nitride, 99.6% (metals basis), Thermo Scientific Chemicals

Catalog number: 041946.18
50 g, Each
Quantity
Catalog number: 041946.18
also known as 041946-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12057-71-5
IUPAC Name
diazenediide; trimagnesium-1,3-bis(ylium)
Molecular Formula
Mg3N2
InChI Key
VZPQZBKNAUHPOS-UHFFFAOYSA-N
SMILES
[N-]=[N-].[Mg+][Mg][Mg+]
Total Metal Impurities
0.4% max.

Description

The use of magnesium nitride as a convenient source of ammonia allows a direct transformation of esters to primary amides. Methyl, ethyl, isopropyl, and tert-butyl esters are converted to the corresponding carboxamides in good yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text